H9890
HA-Ultrogel®
40% (by weight), aqueous ethanol suspension, 60-180 μm
Synonym(s):
Hydroxyapatite preparation
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
form
aqueous ethanol suspension
concentration
40% (by weight)
particle size
60-180 μm
pH
4-13
storage temp.
2-8°C
Application
HA ultrogel is used in affinity chromatography, protein chromatography and adsorption/partition.
Other Notes
Microcrystalline hydroxyapatite in cross-linked 4% beaded agarose
Physical form
Suspension in 1 M NaCl containing 20% ethanol
Legal Information
Ultrogel is a registered trademark of Pall Corporation
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F
Flash Point(C)
40 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Borowski et al.
The Biochemical journal, 331 ( Pt 2), 649-657 (1998-06-11)
The catalytic domain of p72(syk) kinase (CDp72(syk)) was purified from a 30000 g particulate fraction of rat spleen. The purification procedure employed sequential chromatography on columns of DEAE-Sephacel and Superdex-200, and elution from HA-Ultrogel by chloride. The analysis of the
H S Lee et al.
Plant physiology, 112(4), 1513-1522 (1996-12-01)
Neutral and alkaline invertase were identified in cells of a suspension culture of carrot (Daucus carota L.) and purified to electrophoretic homogeneity. Neutral invertase is an octamer with a molecular mass of 456 kD and subunits of 57 kD, whereas
Martha Contreras-Zentella et al.
The Journal of biological chemistry, 278(34), 31473-31478 (2003-06-14)
A novel bo3-type quinol oxidase was highly purified from Bacillus cereus PYM1, a spontaneous mutant unable to synthesize heme A and therefore spectroscopically detectable cytochromes aa3 and caa3. The purified enzyme contained 12.4 nmol of heme O and 11.5 nmol
R K Achazi et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 121(1-3), 339-350 (1999-02-11)
The cytochrome P450 system of the oligochaetes Eisenia f. fetida (tiger worm) and Enchytraeus crypticus (pot worm) was analysed using ethoxy-, pentoxy- and benzoxyresorufin as substrates for monooxygenase activity. Whole body microsomes of the earthworm E.f. fetida displayed PentROD activity
Purification and characterization of the acid lipase from the endosperm of castor oil seeds.
Fuchs, C., et al.
Journal of Plant Physiology, 149(1-2), 23-29 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service