Skip to Content
Merck
All Photos(1)

Key Documents

H7009

Sigma-Aldrich

3-Hydroxy-4,5-dimethoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O5
CAS Number:
Molecular Weight:
198.17
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

COc1cc(cc(O)c1OC)C(O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service