H6028
4′-Hydroxymethyltrioxsalen
Synonym(s):
4′-Hydroxymethyl-4,5′,8′-trimethylpsoralen
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H14O4
CAS Number:
Molecular Weight:
258.27
MDL number:
UNSPSC Code:
12352200
storage temp.
2-8°C
SMILES string
CC1=CC(=O)Oc2c(C)c3oc(C)c(CO)c3cc12
Biochem/physiol Actions
A photosensitizing agent known to intercalate into double stranded DNA. May be useful for targeted mutagenesis and in cross-linking DNA-binding protein to DNA.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S S Sastry et al.
Biochemistry, 32(21), 5526-5538 (1993-06-01)
We have developed a technique for cross-linking DNA binding proteins to DNA using psoralen furanside monoadducts as photoaffinity probes and a continuous-wave argon ion laser (366 nm) as a light source. Several DNA binding proteins (T7 RNA polymerase, UvrB, single-stranded
P A Havre et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(16), 7879-7883 (1993-08-15)
Oligonucleotides can bind as third strands of DNA in a sequence-specific manner in the major groove in homopurine/homopyrimidine stretches in duplex DNA. Here we use a 10-base triplex-forming oligonucleotide linked to a psoralen derivative at its 5' end to achieve
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service