Skip to Content
Merck

H177

2-Hydroxytetradecanoic acid

powder

Synonym(s):

2-Hydroxymyristic acid, DL-α-Hydroxymyristic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H28O3
CAS Number:
Molecular Weight:
244.37
EC Number:
219-714-0
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
1725369
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

color

white

solubility

DMSO: 4 mg/mL (with heating in a 70°C water bath), H2O: 5 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCC(O)C(O)=O

InChI

1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-12-13(15)14(16)17/h13,15H,2-12H2,1H3,(H,16,17)

InChI key

JYZJYKOZGGEXSX-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library