impurities
≤5% Protein (Lowry)
intrinsic viscosity
≥60
suitability
suitable for substrate for hyaluronidase
storage temp.
−20°C
Analysis Note
Also contains chondroitin sulfate.
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mark E Hart et al.
International journal of microbiology, 2009, 614371-614371 (2010-02-05)
Hyaluronidases degrade hyaluronic acid, a major polysaccharide of the extracellular matrix of tissues, and are considered important for virulence in a number of Gram-positive and -negative bacteria. The purpose of the present study was to determine the prevalence of hyaluronidase
Carmela Ricciardelli et al.
BMC cancer, 13, 476-476 (2013-10-16)
Hyaluronan (HA) an important component of the extracellular matrix, has been linked to tumor progression and drug resistance in several malignancies. However, limited data is available for ovarian cancer. This study investigated the role of hyaluronan (HA) and a potential
Yifei Dong et al.
Scientific reports, 6, 36928-36928 (2016-11-22)
The extracellular matrix glycosaminoglycan, hyaluronan, has been described as a regulator of tissue inflammation, with hyaluronan fragments reported to stimulate innate immune cells. High molecular mass hyaluronan is normally present in tissues, but upon inflammation lower molecular mass fragments are