GN683277
Greiner Sapphire PCR tubes, thin wall
0.2 mL, violet, flat cap
Synonym(s):
Greiner PCR tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
material
polypropylene
packaging
pack of 1000 ea
manufacturer/tradename
Greiner 683277
availability
available only in China
technique(s)
PCR: suitable
suitability
suitable for (PCR applications)
Looking for similar products? Visit Product Comparison Guide
General description
Greiner PCR tubes′ thin wall construction promotes optimal heat transfer between the thermocycler block and the reaction mixture. These PCR tubes are certified RNase/DNase free and are non-pyrogenic as well.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service