Skip to Content
Merck
All Photos(1)

Key Documents

GEPA35000

Sigma-Aldrich

Cy®5 Ab Kit

Cytiva PA35000, pack of 1 kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CZK 17,400.00

CZK 17,400.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
CZK 17,400.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CZK 17,400.00


Please contact Customer Service for Availability

Request a Bulk Order

feature

wetted part: no

packaging

pack of 1 kit

manufacturer/tradename

Cytiva PA35000

storage condition

protect from light

color

Blue

solubility

cold water: soluble (Coupling buffer: Soluble in cold water and hot water)
cold water: soluble (Cy5 dye: Soluble in cold water and hot water)
cold water: soluble (Elution buffer: Soluble in cold water and hot water)

storage temp.

2-8°C

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact [email protected].

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service