Skip to Content
Merck
All Photos(2)

Key Documents

GE17-5481-15

HiScreen Capto Core 700

Cytiva 17-5481-15, pkg of 1 column

Synonym(s):

Capto Core 700 Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CZK 5,920.00

CZK 5,920.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
CZK 5,920.00

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

CZK 5,920.00


Please contact Customer Service for Availability

ligand

octylamine

packaging

pkg of 1 column

manufacturer/tradename

Cytiva 17-5481-15

parameter

0.8-3.9 ml/min flow rate
8 bar

bed size

77 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

highly cross-linked agarose

avg. part. size

75 (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

3-13(pH interval where the medium can be subjected to, for cleaning- or sanitization-in-place.)

working range

2-14(pH interval where the medium can be operated without significant change in function.)

capacity

~13 mg binding capacity (ovalbumin)

suitability

suitable for bioprocess medium

General description

Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules. The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography resin.

Capto Core 700 is composed of a ligand-activated core and inactive shell. The inactive shell excludes large molecules (cut off ~ Mr 700 000 [700 kDa]) from entering the core through the pores of the shell. These larger molecules are collected in the column flowthrough while smaller impurities bind to the internalized ligands. The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core. The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations. Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.
Dynamic binding capacities were measured at 10% breakthrough with a residence time of 3 min (1.6 ml/min = 200 cm/h) in HiScreen columns. Buffer: 20 mM Tris-HCl, 150 mM NaCl, pH 7.5.

Features and Benefits

  • For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
  • Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough.
  • Significantly improved productivity and higher flow rates compared with gel filtration methods.
  • Straightforward optimization due to flowthrough chromatography and robust performance.
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiScreen is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service