Skip to Content
Merck
All Photos(1)

Documents

GE17-1318-01

DTT

Cytiva 17-1318-01, pack of 1 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pack of 1 g

manufacturer/tradename

Cytiva 17-1318-01

color

White

pH range

0-5.1

solubility

cold water: soluble (Easily soluble in cold and hot water)

storage temp.

2-8°C

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa K Takahashi et al.
Nature communications, 9(1), 3347-3347 (2018-08-23)
There is a need for large-scale, longitudinal studies to determine the mechanisms by which the gut microbiome and its interactions with the host affect human health and disease. Current methods for profiling the microbiome typically utilize next-generation sequencing applications that
Christoph Kreer et al.
Cell, 182(4), 843-854 (2020-07-17)
The SARS-CoV-2 pandemic has unprecedented implications for public health, social life, and the world economy. Because approved drugs and vaccines are limited or not available, new options for COVID-19 treatment and prevention are in high demand. To identify SARS-CoV-2-neutralizing antibodies
Philipp Schommers et al.
Cell, 180(3), 471-489 (2020-02-01)
Broadly neutralizing antibodies (bNAbs) represent a promising approach to prevent and treat HIV-1 infection. However, viral escape through mutation of the HIV-1 envelope glycoprotein (Env) limits clinical applications. Here we describe 1-18, a new VH1-46-encoded CD4 binding site (CD4bs) bNAb
Kevin A Bockerstett et al.
International journal of molecular sciences, 19(4) (2018-04-13)
The ability to analyze individual epithelial cells in the gastric mucosa would provide important insight into gastric disease, including chronic gastritis and progression to gastric cancer. However, the successful isolation of viable gastric epithelial cells (parietal cells, neck cells, chief

Articles

This page describes protein precipitation as an optional step in sample preparation for 2-D electrophoresis with Cytiva products.

This page describes components of rehydration solutions for use in 2-D Electrophoresis with Cytiva products.

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

This page describes two-dimensional separation of protein samples with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service