Skip to Content
Merck
All Photos(6)

Documents

GE17-1195-01

Sigma-Aldrich

HiPrep 26/60 Sephacryl® S-200 HR

Cytiva 17-1195-01, column L × I.D. 60 cm × 26 mm, 50 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

polypropylene (PP)

mol wt

5000-250000 Da (globular proteins)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 17-1195-01

parameter

<13 mL sample volume
<2.6 mL/min flow rate
0.15 Mpa (22 psi)

bed volume

320 mL

column L × I.D.

60 cm × 26 mm

matrix

dextran-acrylamide particle platform
allyl dextran and N, N’-methylenebisacrylamide
polymeric

matrix active group

dextran-acrylamide phase

avg. part. size

50 μm

cleaning

2-13

working range

3-11

separation technique

size exclusion (SEC)

General description

HiPrep 16/60 Sephacryl® S-200 HR are prepacked gel filtration column for preparative separation of biomolecules from up to 13 mL sample volume. Sephacryl® S-200 is used for separating mid-size proteins including antibodies and serum proteins.

Features and Benefits

  • Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix.
  • Gel filtration columns for low to medium pressure AKTA systems.
  • Well-established in industrial production.
  • The HiPrep Sephacryl® series offers reproducible separations over a wide molecular weight range.
  • Convenient, prepacked HiPrep Sephacryl® columns speed up protein purification.

Suitability

All commonly used buffers, 0.2 M NaOH, 0.1 M HCl, 1 M acetic acid, 8 M urea, 6 M guanidine HCl, 1 % SDS, 20 % ethanol, 30 % propanol, 30 % acetonitrile (tested at 40oC for 1 week), 0.5 M NaOH (only for cleaning-in-place)
Gel filtration columns for low to medium pressure ÄKTA systems.
for preparative separation of biomolecules of up to 13 mL sample volume

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

HiPrep is a trademark of Cytiva
Sephacryl is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page shows how to refold proteins using gel filtration and products from Cytiva.

Sephacryl column separates using Size Exclusion Chromatography media, prepared by covalently cross-linking allyl dextran.

Related Content

Separation options for Size Exclusion Chromatography with Sephacryl from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service