Skip to Content
Merck
All Photos(1)

Key Documents

GE10600124

Amersham Protran® western blotting membrane for use with Minifold I blot-spot system

nitrocellulose membrane, pore size 0.45 μm, W × L 102 mm × 133 mm, sheets, pack of 10 ea

Synonym(s):

blotting and detection, blotting membrane

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CZK 3,310.00

CZK 3,310.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
CZK 3,310.00

About This Item

UNSPSC Code:
41105339
NACRES:
NC.01

CZK 3,310.00


Please contact Customer Service for Availability

material

nitrocellulose membrane
sheets

packaging

pack of 10 ea

manufacturer/tradename

Cytiva 10600124
Cytiva Article No. 29-0912-53 (US reference)

W × L

102 mm × 133 mm

pore size

0.45 μm pore size

capacity

115-125 μg binding capacity(IgG/cm2)

Looking for similar products? Visit Product Comparison Guide

General description

Nitrocellulose membrane suitable for proteins of a wide range of molecular weights. Low background and excellent signal-to-noise ratios, compatible with a variety of labeling and detection systems, high binding capacicity and proprietary nitrocellulose formula gives a long shelf life of bound proteins

Amersham Protran offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems.

Features and Benefits

  • Low background and excellent signal-to-noise ratios
  • Compatible with a variety of labelling and detection systems
  • High binding capacity
  • Suitable for proteins with a wide range of molecular weights
  • Ideal tool for your antibody purification and analysis workflow
  • Proprietary nitrocellulose formula ensures a long shelf life of bound proteins

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service