GBL666215
Grace Bio-Labs Press-To-Seal silicone isolator, PSA both sides
wells, 8 (Square, 7mm x 7mm x 1.8mm depth), (Adhesive on both sides)
Synonym(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
Recommended Products
material
red silicon
feature
wells 8 (Square, 7mm x 7mm x 1.8mm depth)
(Adhesive on both sides)
packaging
pack of 25 ea
manufacturer/tradename
Grace Bio-Labs 666215
external L × W
43 mm × 26 mm
well area
0.38 cm2
wells
8
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service