Documents
GBL665273
Grace Bio-Labs CultureWell silicone isolator sheet material
thickness 1.5 mm, (Adhesive on one side)
Synonym(s):
silicone sheet with single side adhesive
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
material
clear silicon
sterility
non-sterile
feature
(Adhesive on one side)
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 665273
L × W
180 mm × 130 mm
thickness
1.5 mm
General description
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.