All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H20N2O5
CAS Number:
Molecular Weight:
260.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
storage temp.
2-8°C
SMILES string
OC([C@@H](N)CCC(N[C@H](C(O)=O)CC(C)C)=O)=O
InChI
1S/C11H20N2O5/c1-6(2)5-8(11(17)18)13-9(14)4-3-7(12)10(15)16/h6-8H,3-5,12H2,1-2H3,(H,13,14)(H,15,16)(H,17,18)
InChI key
MYFMARDICOWMQP-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Andreas Dunkel et al.
Journal of agricultural and food chemistry, 55(16), 6712-6719 (2007-07-10)
Addition of a nearly tasteless aqueous extract isolated from beans (Phaseolus vulgaris L.) to a model chicken broth enhanced its mouthfulness and complexity and induced a much more long-lasting savory taste sensation on the tongue. Gel permeation chromatography and hydrophilic
C J Li et al.
Phytochemistry, 42(2), 443-445 (1996-05-01)
Our chemical studies on Cycas circinalis seeds from Guam has provided two new nonprotein amino acids, N-(3'-one-5'-methyl)-hexylalanine and leucine betaine. N-methylisoleucine, previously reported as a component of naturally occurring peptides, has been isolated as a free amino acid from the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service