Skip to Content
Merck
All Photos(1)

Documents

G4627

Sigma-Aldrich

Glyoxylic acid monohydrate

≥97%

Synonym(s):

Formylformic acid, Oxoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCO2H · H2O
CAS Number:
Molecular Weight:
92.05
Beilstein:
969535
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥97%

bp

100 °C (lit.)

mp

49-52 °C (lit.)

storage temp.

−20°C

SMILES string

[H]O[H].[H]C(=O)C(O)=O

InChI

1S/C2H2O3.H2O/c3-1-2(4)5;/h1H,(H,4,5);1H2

InChI key

MOOYVEVEDVVKGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 52, 321-331 (2018-07-22)
The purpose of present study was to investigate the applicability of reconstructed human epidermis model to identify skin corrosive UN GHS Categories 1A, 1B/1C and non-corrosive chemicals in China. By using a commercialized reconstructed human epidermis model, China EpiSkin™ which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service