Skip to Content
Merck
All Photos(1)

Key Documents

G3892

Sigma-Aldrich

Gly-Arg-Gly-Asp

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25N7O7
CAS Number:
Molecular Weight:
403.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

NCC(=O)NC(CCCNC(N)=N)C(=O)NCC(=O)NC(CC(O)=O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Tetrapeptide overlapping the cell-attachment domain of fibronectin (the cell-attachment promoting activity has been localized to the tetrapeptide Arg-Gly-Asp-Ser).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignacy Z Siemion et al.
Peptides, 24(8), 1109-1115 (2003-11-13)
Continuing our research on Mycobacteria kansasii phagocytosis inhibition, we have examined in that context three series of peptides derived from the RGDVY and GRGD sequences. It was found that the levels of the inhibitory activity depend on the amino acid
Tze-Wen Chung et al.
Biomaterials, 23(24), 4803-4809 (2002-10-04)
To improve ECs adhesion and growth on chitosan, cell adhesive peptide Gly-Arg-Gly-Asp (GRGD) was photochemically grafted to its surface. Grafting 0.025 M of GRGD-SANPAH (N-Succinimidyl-6-[4'-azido-2'-nitrophenylamino]-hexanoate) solutions to chitosan and tripolyphosphate anhydrous crosslinked chitosan (chitosan-TPP) surfaces was performed by surface adsorption
Sidi A Bencherif et al.
Biomacromolecules, 10(7), 1795-1803 (2009-06-13)
This study presents the synthesis and evaluation of cell adhesive poly(ethylene oxide) (PEO) star polymers for potential biomedical applications. Star polymers with a size of approximately 20 nm and with relatively low polydispersities (M(w)/M(n) ≤ 1.6), containing GRGDS (Gly-Arg-Gly-Asp-Ser) segments
Shoei-Shen Wang et al.
Journal of biomedical materials research. Part A, 91(3), 753-761 (2008-12-04)
Accelerated thrombolysis by pressure-driven permeation has been demonstrated in in vitro and in vivo animal models by using plasminogen activators (PAs) encapsulated liposomes or PEG microparticles. Recent reports have also described acceleration of thrombolysis using tissue type PA (t-PA) encapsulated
Y S Lin et al.
Artificial organs, 25(8), 617-621 (2001-09-05)
To improve endothelial cell adhesion and growth on the surface of polyethylene glycol modified polyurethane (PU-PEG), cell adhesive peptide Gly-Arg-Gly-Asp (GRGD) was photochemically grafted to the surface. The surface grafted GRGD-N-Succinimidyl-6-[4'-azido-2'-nitrophenylamino]hexanoate (SANPAH) on a PU-PEG surface was performed by adsorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service