Skip to Content
Merck

F4229

Monoclonal Anti-Fas (CD95/Apo-1)−FITC antibody produced in mouse

clone DX2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Apo-1, Anti-CD95, Monoclonal Anti-Fas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DX2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 1:50 using Cultured human Burkitt′s lymphoma Raji cells

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... FAS(355)

General description

Many cells can be activated to undergo apoptosis following the interaction of selected ligands with cell surface receptors. The most well studied receptors are CD95/Fas/Apo-1 (apoptosis inducing protein 1) and tumor necrosis factor receptor 1 (TNFR1). Apoptosis mediated by either of these results in activation of the caspases. However, Fas-mediated death occurs much more rapidly than that triggered by TNFR1. Human Fas/CD95/Apo-1 is a single transmembrane glycoprotein receptor (325 amino acids, 45-48 kDa).

Immunogen

murine L cells transfected with a human Fas/CD95 cDNA.

Application

Monoclonal Anti-Fas (CD95/Apo-1)-FITC antibody is suitable for flow cytometry at a dilution of 1:50 using cultured human Burkitt′s lymphoma Raji cells.

Biochem/physiol Actions

APO-1/Fas(CD95) comprises of a death domain (DD) within the cytoplasmic region which triggers apoptosis upon binding of their cognate ligands. Once it is activated, APO-1/Fas(CD95) further aggregates its intracellular death domains which leads to the recruitment of two key signaling proteins followed by the formation of death-inducing signaling complex. These complex crosslinks through its C-terminal DD with APO-1/Fas receptors and engage caspase-8 via its N-terminal death effector domain (DED) to the DISC.
Reacts specifically with the functional epitope of human Fas (CD95/Apo-1) antigen. By immunoblotting, the clone recognizes denatured, non-reduced recombinant human Fas (amino acid residues 1-173). The antibody is reactive in flow cytometry, and may be reactive in the induction of apoptosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library