Skip to Content
Merck

F1028

Farnesyl pyrophosphate-[1-3H(N)] triammonium salt

aqueous ethanol solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H37N3O7P2
CAS Number:
Molecular Weight:
433.42
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous ethanol solution

extent of labeling

15-30 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

N.[H]C(OP(O)(=O)OP(O)(O)=O)\C=C(/C)CC\C=C(/C)CC\C=C(\C)C

Packaging

Packaged under Nitrogen

Physical form

Ethanol: water (1:1) containing 0.25 M ammonium bicarbonate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library