Product Name
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Quality Segment
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
L-glutamine: no
NaHCO3: yes
HEPES: no
glucose: low
phenol red: yes
sodium pyruvate: yes
shipped in
ambient
storage temp.
2-8°C
General description
This modification has been shown to improve the stability of the medium.
Preparation Note
Supplement with 0.584 g/L glutamine.
Still not finding the right product?
Explore all of our products under Dulbecco′s Modified Eagle′s Medium - low glucose
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
