Skip to Content

F0166

Factor XIIIa Fragment 72-97

≥90% (HPLC)

Synonym(s):

Asn-Lys-Leu-Ile-Val-Arg-Arg-Gly-Gln-Ser-Phe-Tyr-Val-Gln-Ile-Asp-Phe-Ser-Arg-Pro-Tyr-Asp-Pro-Arg-Arg-Asp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C144H225N45O40
CAS Number:
Molecular Weight:
3226.60
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CC(N)=O)[C@@H](C)CC)C(C)C)C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N4CCC[C@H]4C(=O)N[C@@H](Cc5ccc(O)cc5)C(=O)N[C@@H](CC(O)=O)C(=O)N6CCC[C@H]6C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(O)=O)C(O)=O

Gene Information

human ... F13A1(2162)

Biochem/physiol Actions

Inhibits the transglutaminase activity of Factor XIIIa, thus blocking the cross-linking of fibrin, fibronectin, and N,N′-dimethylcasein.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library