Skip to Content
Merck

Skip To

EPI024

Core Histone Isolation Kit

sufficient for 100 extractions

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.25
UNSPSC Code:
41116158


usage

sufficient for 100 extractions

storage temp.

−20°C

General description

Histones are small alkaline proteins that interact with eukaryotic DNA to form the fundamental subunit of chromatin known as the nucleosome. The core histones H2A, H2B, H3 and H4 form the central spooling unit of the nucleosome; with two copies of each comprising a histone octamer that interacts with the negatively charged DNA. They enable an ordered compaction of the DNA, as well as spatiotemporal control of gene expression and genomic replication throughout mitosis. Post-translational modification of this core complex results in structural changes within the chromatin and epigenetic control of gene expression. 

Application

Suitable for the isolation of intact functional core histones from tissue and cultured cells, protein profiling, post-translational modification and epigenetic analyses and Western blot and ELISA

Biochem/physiol Actions

The Core Histone Isolation Kit offers an optimized suite of reagents to rapidly extract the total core histones, while preserving acetylation from tissues and cultured cells for biochemical and epigenetic analyses. One hundred standard extractions at a scale of 107 cells or 100 mg of tissue can be performed with the reagents in this kit. Yield of the total histone proteins is ~0.4 mg per 107 cells or 100 mg of tissue.


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters