material
polypropylene bottle
polypropylene cap
round bottle
feature
graduations: no
with cap
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5810729009
capacity
400 mL
General description
400 mL wide-neck bottle which are plastic vessels for preparing, transporting, storing and centrifuging solid and liquid samples and reagents. When centrifuging at high rotational speeds or high temperatures, push the supplied sleeve over the wide-neck bottle. The 400 mL wide-neck bottle does not fit under the cap of the rectangular bucket for therotor A-4-81. It is autoclavable (121C, 20 mins).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG