EP5804774000
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 1 tube 50 ml, pack of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
holds 1 tube 50 ml
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804774000
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-34-6-38
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service