EP4861000813
Eppendorf® Xplorer® Plus 12-channel pipettor
volume 15-300 Å3, multifunction rocker switch, orange, for pipette tips 300 ul
Synonym(s):
Eppendorf 12-Channel Pipette
Select a Size
CZK 39,100.00
Select a Size
About This Item
CZK 39,100.00
Recommended Products
feature
for pipette tips 300 ul
multifunction rocker switch
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 4861000813
volume
15-300 Å3
color
orange
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | EP4861000791 | EP4861000775 | EP4861000830 |
---|---|---|---|
feature for pipette tips 300 ul, multifunction rocker switch | feature for pipette tips 200 ul, multifunction rocker switch | feature for pipette tips 20 ul, multifunction rocker switch | feature for pipette tips 1250 ul, multifunction rocker switch |
packaging case of 1 ea | packaging case of 1 ea | packaging case of 1 ea | packaging case of 1 ea |
manufacturer/tradename Eppendorf® 4861000813 | manufacturer/tradename Eppendorf® 4861000791 | manufacturer/tradename Eppendorf® 4861000775 | manufacturer/tradename Eppendorf® 4861000830 |
color orange | color yellow | color medium gray | color green |
General description
- Multi-functional rocker with up is up and down is down functionality
- Selection dial for quick and easy selection of functions
- Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
- Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
- Powerful rechargeable battery for up to 8 hours work without the need for recharging
- Separate power socket: Work can continue while charging
- Adjustment to external conditions (e.g. geographic height, liquids with high density)
- Lower part of pipette can be autoclaved to ensure decontamination
- 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
- Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service