Documents
EP3880000011
Eppendorf® IsoTherm-System starter set
for 0.5 mL tubes, pkg of 1 ea
About This Item
UNSPSC Code:
41102404
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3880000011
Features and Benefits
  • Eppendorf® IsoTherm-System starter set, includes IsoRack (work rack), IsoSafe (insulating box), and two IsoPacks (cool packs).
  • Ideal for cooling, transporting, and storing deep-frozen samples.
  • Prevents enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
  • Dry incubation technology reduces contamination risk associated with classical icebath incubation for safe sample handling.
  • Holds up to 24 micro tubes for broad flexibility.
  • Two versions of IsoPack (cold pack) available: White maintains 0 °C for up to 6 hours. Blue maintains -21 °C for up to 3 hour.
  • Racks are stackable, autoclavable and can be centrifuged in the MTP rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG