Skip to Content
Merck
All Photos(1)

Key Documents

EP30129555

Eppendorf twin.tec® PCR Plates 96

skirted, green, pkg of 300 ea

Sign Into View Organizational & Contract Pricing

Select a Size

300 EA
CZK 38,900.00

CZK 38,900.00


Please contact Customer Service for Availability


Select a Size

Change View
300 EA
CZK 38,900.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
green wells
size:
96 well
binding type:
low binding surface
feature:
skirt
skirted

CZK 38,900.00


Please contact Customer Service for Availability

material

green wells

feature

skirt
skirted

packaging

pkg of 300 ea

manufacturer/tradename

Eppendorf® 30129555

technique(s)

PCR: suitable

size

96 well

color

green

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.

Application

PCR

Features and Benefits

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service