EP1201
Sigma-Aldrich® MSCHOICETRIO horizontal gel electrophoresis system
run up to 210 samples with 3 tray options
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
Recommended Products
usage
Up to 140 samples for 17 x 10 cm tray
Up to 210 samples for 15 x 15 cm tray
Up to 70 samples for 15 x 7 cm tray
W × L × H
26.5 cm × 17.5 cm × 9.0 cm
buffer volume
500 mL
Application
Includes a 15 x 7 cm, 15 x 10 cm, and 15 x 15 cm UV Trays, 2 x 20 sample combs, loading guides and dams.
Features and Benefits
This unit features low buffer volumes and can run up to 210 samples with 3 tray options. Multichannel pipette compaticle combs for speed loading not included.
Components
Corrosion resistant gold plated lid terminals with retractable connectors compatible with most major power supplies.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service