Skip to Content
Merck
All Photos(1)

Documents

EP022364120

Eppendorf® Flex-Tubes Microcentrifuge Tubes

volume 1.5 mL, colorless, suitable for PCR, pack of 500 tubes

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703

material

colorless
polypropylene

sterility

(PCR clean)

feature

graduated
Flex-Tube lid

packaging

pack of 500 tubes

manufacturer/tradename

Eppendorf® 022364120 (NA)

technique(s)

PCR: suitable

volume

1.5 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The easy-open, easy-close design of the Flex-Tube provides convenient, ergonomic, and reliable one-handed operation. Whether sample preparation, centrifugation or storage – the Flex-Tube fit perfectly into your workflows. This is the successor to the renowned first microcentrifuge tube 3810 from Eppendorf. The idea of manufacturing a 1.5 mL plastic test tube was born out of the necessity to introduce an appropriate container into the automatic clinical analyzers developed by Eppendorf in 1962. The tried and tested design with invaluable features shows perfect sealing qualities, excellent chemical stability and optimal construction materials for resisting the demands of centrifugal forces. As they are easy to open and close securely, The Eppendorf Flex-Tubes off er comfort and dependability for sample preparation, centrifugation and storage. One vessel for all applications in molecular biology, chemistry and bioche mistry – safer than ever. The Eppendorf Flex-Tubes can be centrifuged up to 30,000 x g without any problems.

Application

Eppendorf™Flex-Tubes Microcentrifuge Tubes are ideal for preparing samples, storage, or centrifugation

Features and Benefits

  • Easy to open and close tube lid for ergonomic use
  • Made up of polypropylene which provides better mechanical, chemical, and temperature resistance
  • Sample loss can be minimized due to tube breakage and ensures safety while working with dangerous samples
  • Plasticizers, slip agents, and biocides were not used in the manufacturing process
  • Graduated with frosted lid for easy labeling
  • When open it can be autoclavable
  • Functionality is guaranteed from -86 C to 100 C
  • Simple verifi cation of the pipetted volume by clear graduation
  • All variants can be autoclaved for subsequent use in sterile environments (121 C, 20 min.)
  • Manufactured without the use of slip agents, plasticizers and biocides
  • High resistance to chemicals, mechanical stress and temperature extremes – manufactured from virgin polypropylene
  • Tube lid is easy to open and close for ergonomic handling
  • Easy labeling on frosted lid and surface on the side

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service