Skip to Content
Merck
All Photos(1)

Documents

EP0030602200

Eppendorf® Microplate 96/U-PP, 96 wells

white polypropylene, round clear wells, sterile, pkg of 80 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

round clear wells
white polypropylene

sterility

sterile; autoclaved
sterile

packaging

pkg of 80 ea

manufacturer/tradename

Eppendorf® 0030602200

technique(s)

PCR: suitable

size

96 wells

Looking for similar products? Visit Product Comparison Guide

General description

  • The Eppendorf Microplates bring an outstanding clarity to your laboratory.
  • It has never been this easy to pipette, control and recover samples with a polypropylene microplate.
  • The easy-to-read alphanumeric label of the OptiTrack® matrix on each plate makes it unbelievably easy to identify individual wells, speeds up the filling process and significantly reduces the risk of errors.
  • The high transparency is outstanding and ensures that you can always quickly locate your samples.
  • The Microplates are also available in Eppendorf LoBind material for maximum sample recovery when only limited amount of sample material is available.

Pipette, check and recover your samples easily with Eppendorf round bottom Microplates. The easy-to-read OptiTrack alphanumeric matrix label for identifying each well speeds up filling and significantly reduces the risk of pipetting errors.

Application

  • Sample storage and preparation.
  • Assays that require high resistance against temperature or solvents.
  • Active ingredient screening.
  • Combinatorial chemistry.
  • Storage of cDNA or genomic banks.
  • Protein analysis, Genotyping.
  • Storage of siRNA or oligonucleotide libraries.
  • Preparation of tissue and plant lysates.

  • sample storage and preparationassays that require high resistance against temperature or solvents
  • active ingredient screening
  • combinatorial chemistry
  • storage of cDNA or genomic banks
  • protein analysis
  • genotyping
  • storage of siRNA or oligonucleotide libraries
  • preparation of tissue and plant lysates

Features and Benefits

  • Unmatched, transparent polypropylene (PP) for improved sample visibility.
  • OptiTrack® Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
  • RecoverMax® well design: Optimized well geometry for minimal remaining volume and excellent mixing properties.
  • Raised well rims and smooth surface for reliable closing, even with heat sealing.
  • g-Safe®: exceptional centrifugation stability up to 6,000 × g.
  • Polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without slip agents, plastocizers or biocides − Materials which could have a negative effect on bioassays.
  • Guaranteed functionality from -86 °C to 100 °C.
  • Autoclavable (121 °C, 20 min).

  • Eppendorf Microplates consist of polypropylene wells which enhance sample visibility. In addition, polypropylene also ensures resistance towards chemicals, mechanical stressors and high temperatures.
  • A raised well rim and a smooth surface facilitates secure sealing.
  • Eppendorf Microplates can withstand centrifugal forces up to six thousand times the gravitational force.
  • The OptiTrack matrix feature enables faster well identification and reduces pipetting error.
  • An optimized RecoverMax well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • Eppendorf Microplates function efficiently over a wide temperature range, ranging from -86 °C to 100 °C.
  • These microplates can be autoclaved at 121 °C for 20 min.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service