Skip to Content
Merck
All Photos(1)

Key Documents

EP0030108531

Eppendorf® Protein LoBind tubes

new

capacity 25 mL, PCR clean, cap (snapTec), speed 14,000 x g rpm, pkg of 200 ea (5 bags x 40 tubes)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
Pricing and availability is not currently available.

material

cap (snapTec)
conical bottom
polypropylene

grade

for proteomics

description

withstand 17,000xg

sterility

non-sterile

feature

DNase free
PCR clean
RNase free
graduations: no
lid

packaging

pkg of 200 ea (5 bags x 40 tubes)

manufacturer/tradename

Eppendorf® 0030108531

parameter

14,000 x g rpm speed

capacity

25 mL

color

colorless

suitability

suitable for PCR
suitable for molecular biology

binding type

Protein low binding

Looking for similar products? Visit Product Comparison Guide

General description

Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Protein LoBind Tubes, SnapTec cap, Protein LoBind, 25 mL, PCR clean, colorless, 200 tubes (5 bags × 40 tubes)

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing to minimize evaporation
  • Rated up to 14,000_x_g centrifugation speed
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service