Skip to Content
Merck
All Photos(1)

Documents

EP0030089936

Eppendorf® Combitips advanced assortment pack

contains 1 Combitp ad- vanced of each size and one 25 mL and one 50 mL adapter

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

feature

DNase free

packaging

pack of 1 (1 x each size 25 / 50 mL)

manufacturer/tradename

Eppendorf® 0030089936

General description

Combitips advanced assortment pack, contains 1 Combitp ad- vanced of each size and one 25 mL and one 50 mL adapter

Application

LIQUID HANDLING

Features and Benefits

contains one Combitips® advanced sample of each size and Adapter advanced 25 and 50 mL
Eppendorf Quality

Legal Information

Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service