Skip to Content
Merck

Skip To

EMU190891

MISSION® esiRNA

targeting mouse Urb2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
CZK 5,620.00
50 μg
Please contact Customer Service for Availability
CZK 10,100.00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

CZK 5,620.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCAGTATTATTCTCTGGGGATCTCCCTAGCCAGAGCTGCTCTGGATAGTTCAAACCTCCCTTTGTTGCTACCAGGTGTAGAAATGGAGTTTTGGAAGAAGGTGGAAGAGTGTGTAGGCCAGTCCAGGTCTCTCAGTAGGTACTGCTTAGAACAGCTGTATCTTCAGAAGGTGAAAAGGACTTTAATGCGGAGTAATTCACAATCCAAAGAAGCCCTTCAAACCTTGAGGTTTGACACTGCTCACATTCTTGATTCTAGCAGAGACTGTTTAAGTCAGAAGACAGCAGCTGCCTGGGATAGGCAGGTGTCGACAATGAATGAGTCCATATATCCTGTAGCACACTGGCACTTGATTGTATCAAACCTCACAGTTTTAATACCCTACCTTTGTTTGAATGATGTGAGATATGTGGCCACTGTCTTGTTAAGAACTTTACCAGCAAGTAAAGCCCAGGGAAGCTTGGCACATGGTGAGCCACATGTCACACTTGAGAAAATATCCACAGCCCTCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU056651EMU052721EMU066001
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

GCCAGTATTATTCTCTGGGGATCTCCCTAGCCAGAGCTGCTCTGGATAGTTCAAACCTCCCTTTGTTGCTACCAGGTGTAGAAATGGAGTTTTGGAAGAAGGTGGAAGAGTGTGTAGGCCAGTCCAGGTCTCTCAGTAGGTACTGCTTAGAACAGCTGTATCTTCAGAAGGTGAAAAGGACTTTAATGCGGAGTAATTCACAATCCAAAGAAGCCCTTCAAACCTTGAGGTTTGACACTGCTCACATTCTTGATTCTAGCAGAGACTGTTTAAGTCAGAAGACAGCAGCTGCCTGGGATAGGCAGGTGTCGACAATGAATGAGTCCATATATCCTGTAGCACACTGGCACTTGATTGTATCAAACCTCACAGTTTTAATACCCTACCTTTGTTTGAATGATGTGAGATATGTGGCCACTGTCTTGTTAAGAACTTTACCAGCAAGTAAAGCCCAGGGAAGCTTGGCACATGGTGAGCCACATGTCACACTTGAGAAAATATCCACAGCCCTCC

esiRNA cDNA target sequence

CCTTCTCAATGGCTGCATCTCCCAAACTTGCCGCATCTCCCAAACCTGCCACATCTCCCAAGCCTTTGCCCTCACCTAAGCCTTCTGTTTCACCCAAGCCCTCTCTGTCAGCTAAGCCTTCAATATCTACTAAACAGATTTCTAAATCCAACCCAGCTCCCAAGCCTGCTGTATGCCCAAGTTCTTCTAGTCCAAACACACTAGTAGCCCAGAGTAGCCACTCCACAAGTAACAACCCTGTCCATAAACAGCCCAGTGGAATGAACATCAGCAGACAGTCTCCCACTCTGAATTTGTTGCCCTCAAATCGCACTTCTGGCCTTCCGACTACAAAAACTCTTCAGGCCCCTTCTAAACTAACAAACTCATCATCCACTGGAACTGCTGGGAAGAACAGCTTGAGTGGAATCCCAATGAATGTACCTGCCAGCAGAGGTAGCAACCTTAACTCAAGTGGAGCTAATAGGACTAGTCTATCTGGGGGAACAGGAAGTGGAACACAGGG

esiRNA cDNA target sequence

ATCAGTGCGGAGAAGAGGAAAGACAAAGAGAGGCTTGTTGAAATGCTCCGAGCCATGAAGCAGAGAGCACTGTCCGCAGCAGACTCAGTGACAAACTCTTCGAGGGACAGTCCTCCCGTCTCCCTGAGTGAACCAGCCACACAGCCAGCTCCTCTAGAGACGGATCAGCCTGTCGGGGTCCCTGCCTCCTTGTCAGATGTCCCAAAGACCACGGAGACCGGGAGACTGGAACAGCTCCGGCCCCAGGAGCTCTTGAGAGTCCAGGAGCCGGCTCCTCCCAGTGGTGAGAAGGCCAGGCTGAGCGAGGCCCCTGGTGGCAAGAAGAGCCTGAGCATGCTCCATTACCTCCGGGGCGCCGCGCCCAAGGACATTCCTGTGCCGTTGTCTCACAGCATCAACGGGAAGAGCAAGCCCTGGGAGCCTTTCGTGGCCGAAGAGTTTGCACATCAATTCCACG

esiRNA cDNA target sequence

CTTCCGGAGAGTCCTGTGAGAGATGGCTGCAAAGGTGCTCAGGAGGGAATCTGTGGCCGGGTCAAGCTTGGCCTCAGGCCTTCTCTTCTTGTTCATGTTGAAGCTGTCATCCATTCCCCAGACTTCCTCCCAGCCTCTCTGCATGTCCTCAGTGGACCTGTCTTTCGGAAGTGCCACATCTTACCTTTCATGATGCCAGCATTTCACGAGACTTTATTCATCCTTGGATTTGATAACAATATGAAGGAGAGTTGCCTTCCGTCGCTGCTGGGTCACCTGAAGGATGCTCTGGGTAACCTTCTGACTCAGACATTGCAGGAGGGCTCTAGTTTGGGCACTTCGGTGGAATTTGTCCTTCAACCAAACGGAAAGGATTATATAATTCATGTGAAGTCTCTTAATTTTGGCCCAGATTG

Ensembl | mouse accession no.

ENSMUSG00000031976

Ensembl | mouse accession no.

ENSMUSG00000038538

Ensembl | mouse accession no.

ENSMUSG00000031822

Ensembl | mouse accession no.

ENSMUSG00000037845

Gene Information

mouse ... URB2(382038), Urb2(382038)

Gene Information

mouse ... UBN2(320538), Ubn2(320538)

Gene Information

mouse ... GSE1(382034), Gse1(382034)

Gene Information

mouse ... FDXACB1(382137), D630004A14Rik(382137)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_001029876

NCBI accession no.

NM_177185

NCBI accession no.

NP_001139369

NCBI accession no.

NM_198675


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters