Skip to Content
Merck

Skip To

EMU034261

MISSION® esiRNA

targeting mouse Rbks

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 620,00 CZK
50 μg
Please contact Customer Service for Availability
10 100,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

5 620,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTACAGGAACAGCCTCCATAATTGTCAATAATGAAGGCCAGAATATCATCGTCATAGTGGCTGGAGCAAATCTGTTTTTGAATTCTGAAGACTTGAAGAAGGCAGCCAGTGTCATTAGCCGAGCCAAAGTGATGATATGCCAGCTAGAAATAAGCCCGGCAGCTTCTCTGGAAGCTCTGACAATGGCCCGCAGGAGTGGAGTGAAAACCTTGTTCAACCCAGCTCCAGCCATGGCTGACCTGGACCCCCAGTTCTATACCCTCTCCAGTATTTTCTGCTGCAACGAGAGCGAGGCTGAGATTTTAACAGGCCACGCAGTGAGTGACCCCACCACGGCTGGGAAGGCTGCAATGATTCTCCTAGAACGGGGCTGCCAGGTTGTGGTCATCACCTTAGGGGCTTCAGGATGCGTGATACTGTCACAGGCTGAGCCTGTTCCAAAGCACATTCCCACTGAAGCTGTCAAGGCTGTAGACACCACG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU003241EMU034641EMU066341
description

targeting mouse Rbks

description

targeting mouse Rcbtb1

description

targeting mouse Stoml3

description

targeting mouse Spg7

Ensembl | mouse accession no.

ENSMUSG00000029136

Ensembl | mouse accession no.

ENSMUSG00000035469

Ensembl | mouse accession no.

ENSMUSG00000027744

Ensembl | mouse accession no.

ENSMUSG00000000738

esiRNA cDNA target sequence

AGCTACAGGAACAGCCTCCATAATTGTCAATAATGAAGGCCAGAATATCATCGTCATAGTGGCTGGAGCAAATCTGTTTTTGAATTCTGAAGACTTGAAGAAGGCAGCCAGTGTCATTAGCCGAGCCAAAGTGATGATATGCCAGCTAGAAATAAGCCCGGCAGCTTCTCTGGAAGCTCTGACAATGGCCCGCAGGAGTGGAGTGAAAACCTTGTTCAACCCAGCTCCAGCCATGGCTGACCTGGACCCCCAGTTCTATACCCTCTCCAGTATTTTCTGCTGCAACGAGAGCGAGGCTGAGATTTTAACAGGCCACGCAGTGAGTGACCCCACCACGGCTGGGAAGGCTGCAATGATTCTCCTAGAACGGGGCTGCCAGGTTGTGGTCATCACCTTAGGGGCTTCAGGATGCGTGATACTGTCACAGGCTGAGCCTGTTCCAAAGCACATTCCCACTGAAGCTGTCAAGGCTGTAGACACCACG

esiRNA cDNA target sequence

GGCCAATCTTCACTCTGCTCTCACCCCAGGAAGCTGGATCTATCAGGAAAGCATGTGTCTTTGGTACCTCTGCCAATGAGGCGATCTATGTCACTGACAATGATGAGGTCTTCGTGTTTGGACTGAACTACAGTAACTGTCTAGGAACTGGAGATAACCAGAGCACTCTGGTGCCCAAGAAACTAGAAGCTTTGTGTGGAAAAAAGATCAAAAGTCTTAGTTACGGGAGTGGCCCCCATGTCCTCCTCACCACTGAAGATGGAGTCGTCTATGCCTGGGGCCATAATGGATACAGCCAACTGGGAAATGGGACCACCAACCAAGGCATTGCTCCTGTCCAAGTCTGTACTAATCTCTTGATCAAGCAGGTGATTGAAGTAGCTTGTGGTTCACATCATTCGATGG

esiRNA cDNA target sequence

GATTCACCGGAGAAACTGGAGAAGAACAATTTAGTGGGCACCAACAAAAGCCGGCTTGGTGTGTGCGGCTGGATCCTGTTTTTCCTTTCCTTCCTGCTGATGCTCGTTACCTTCCCAATCTCAGTATGGATGTGCTTGAAGATCATTAAGGAGTATGAGCGCGCTGTTGTCTTCAGACTGGGACGCATCCAAGCTGATAAAGCCAAGGGACCAGGTTTGATCCTGGTCCTGCCTTGCATTGACGTGTTTGTCAAAGTAGATCTTCGAACGGTTACTTGTAACATTCCTCCACAAGAGATCCTCACGAGAGACTCTGTGACCACCCAAGTGGATGGAGTCGTCTATTACAGAATCTACAGCGCCGTCTCAGCCGTGGCCAATGTGAACGATGTTCACCAGGCCACGTTCCTGCTGGCTCAGACCACTCTGAGAAATGTCTTAGGGACACAGACCTTGTCCCAGATCTTGTCTGGGCGAGAAGAGAT

esiRNA cDNA target sequence

AAGGCAAGACCCCTGAAGATGATGAAGAAGAGAAGAGACGAAAAGAGCGGGAAGACCAGATGTACCGAGAGAGGCTGCGCACCCTGTTCATCATTGCTCTTGTCATGAGCCTGCTGAACTCCCTCAGTACAAGTGGGGGCAGTACTTCCTGGGCTGACTTTGTCAACGAGATGCTGGCTAAAGGCGAGGTGCAGCGTGTGCAGGTGGTGCCCGAGAGTGATGTGGTGGAAGTCTATCTGCATCCTGGAGCTGTGGTGTTTGGGCGGCCTCGGCTGGCCCTGATGTATCGGATGCAGGTTGCAAACATCGACAAATTTGAAGAGAAGCTTCGAGCAGCCGAAGATGAACTGAACATTGAGAGCAAGGACAGGATTCCCGTGTCCTACAAGCGGACAGGATTCTTTGGGAATGCCCTCT

Gene Information

mouse ... RBKS(71336), Rbks(71336)

Gene Information

mouse ... RCBTB1(71330), Rcbtb1(71330)

Gene Information

mouse ... STOML3(229277), Stoml3(229277)

Gene Information

mouse ... SPG7(234847), Spg7(234847)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

NCBI accession no.

NM_153196

NCBI accession no.

NP_082040

NCBI accession no.

NM_153156

NCBI accession no.

NM_153176

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters