Skip to Content
Merck
All Photos(1)

Key Documents

EHU132681

Sigma-Aldrich

MISSION® esiRNA

targeting human IFI16

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CZK 6,310.00
50 μG
CZK 11,300.00

CZK 6,310.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
CZK 6,310.00
50 μG
CZK 11,300.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CZK 6,310.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGCTCCACCCAACAGTTCTTCAACTGAGAACCCGAAAACAGTGGCCAAATGTCAGGTAACTCCCAGAAGAAATGTTCTCCAAAAACGCCCAGTGATAGTGAAGGTACTGAGTACAACAAAGCCATTTGAATATGAGACCCCAGAAATGGAGAAAAAAATAATGTTTCATGCTACAGTGGCTACACAGACACAGTTCTTCCATGTGAAGGTTTTAAACACCAGCTTGAAGGAGAAATTCAATGGAAAGAAAATCATCATCATATCAGATTATTTGGAATATGATAGTCTCCTAGAGGTCAATGAAGAATCTACTGTATCTGAAGCTGGTCCTAACCAAACGTTTGAGGTTCCAAATAAAATCATCAACAGAGCAAAGGAAACTCTGAAGATTGATATTCTTCACAAACAAGCTTCAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stine Søby et al.
Herpesviridae, 3(1), 6-6 (2012-10-16)
Innate recognition is essential in the antiviral response against infection by herpes simplex virus (HSV). Chemokines are important for control of HSV via recruitment of natural killer cells, T lymphocytes, and antigen-presenting cells. We previously found that early HSV-1-mediated chemokine
A Berry et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(6), 1700-1713 (2010-01-21)
Previously, we used cDNA expression profiling to identify genes associated with glucocorticoid (Gc) sensitivity. We now identify which of these directly influence Gc action. Interferon-inducible protein 16 (IFI16), bone morphogenetic protein receptor type II (BMPRII), and regulator of G-protein signaling
Cindy Orvain et al.
The Journal of clinical investigation, 130(7), 3777-3790 (2020-04-03)
Hidradenitis suppurativa (HS) is a chronic, relapsing, inflammatory skin disease. HS appears to be a primary abnormality in the pilosebaceous-apocrine unit. In this work, we characterized hair follicle stem cells (HFSCs) isolated from HS patients and more precisely the outer
Xin Duan et al.
PloS one, 6(5), e19532-e19532 (2011-05-17)
Glucose restriction in cells increases the AMP/ATP ratio (energetic stress), which activates the AMPK/p53 pathway. Depending upon the energetic stress levels, cells undergo either autophagy or cell death. Given that the activated p53 induces the expression of IFI16 protein, we
Yuanyuan Yang et al.
Hepatology (Baltimore, Md.), 71(4), 1154-1169 (2019-08-14)
Nuclear-located covalently closed circular DNA (cccDNA) of hepatitis B virus (HBV) is a determining factor for HBV persistence and the key obstacle for a cure of chronic hepatitis B. However, it remains unclear whether and how the host immune system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service