Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | 5 620,00 CZK |
50 μg | Please contact Customer Service for Availability | 10 100,00 CZK |
About This Item
5 620,00 CZK
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GGGATGCTTGAAGATGGAAAGAAATTTGATTCCTCCCGGGACAGAAACAAGCCCTTTAAGTTTATGCTAGGCAAGCAGGAGGTGATCCGAGGCTGGGAAGAAGGGGTTGCCCAGATGAGTGTGGGTCAGAGAGCCAAACTGACTATATCTCCAGATTATGCCTATGGTGCCACTGGGCACCCAGGCATCATCCCACCACATGCCACTCTCGTCTTCGATGTGGAGCTTCTAAAACTGGAATGACAGGAATGGCCTCCTCCCTTAGCTCCCTGTTCTTGGATCTGCCATGGAGGGATCTGGTGCCTCCAGACATGTGCACATGAATCCATATGGAGCTTTTCCTGATGTTCCACTCCACTTTGTATAGACATCTGCCCTGACTGAATGTGTTCTGTCACTCAGCTTTGCTTCCGACACCTCTGTTTC
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... FKBP1A(2280)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting human FKBP1A | description targeting human FKBP1B | description targeting human FKBP5 | description targeting human FKBP2 |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence GGGATGCTTGAAGATGGAAAGAAATTTGATTCCTCCCGGGACAGAAACAAGCCCTTTAAGTTTATGCTAGGCAAGCAGGAGGTGATCCGAGGCTGGGAAGAAGGGGTTGCCCAGATGAGTGTGGGTCAGAGAGCCAAACTGACTATATCTCCAGATTATGCCTATGGTGCCACTGGGCACCCAGGCATCATCCCACCACATGCCACTCTCGTCTTCGATGTGGAGCTTCTAAAACTGGAATGACAGGAATGGCCTCCTCCCTTAGCTCCCTGTTCTTGGATCTGCCATGGAGGGATCTGGTGCCTCCAGACATGTGCACATGAATCCATATGGAGCTTTTCCTGATGTTCCACTCCACTTTGTATAGACATCTGCCCTGACTGAATGTGTTCTGTCACTCAGCTTTGCTTCCGACACCTCTGTTTC | esiRNA cDNA target sequence CGTGGAGATCGAGACCATCTCCCCCGGAGACGGAAGGACATTCCCCAAGAAGGGCCAAACGTGTGTGGTGCACTACACAGGAATGCTCCAAAATGGGAAGAAGTTTGATTCATCCAGAGACAGAAACAAACCTTTCAAGTTCAGAATTGGCAAACAGGAAGTCATCAAAGGTTTTGAAGAGGGTGCAGCCCAGATGAGCTTGGGGCAGAGGGCGAAGCTGACCTGCACCCCTGATGTGGCATATGGAGCCACGGGCCACCCCGGTGTCATCCCTCCCAATGCCACCCTCATCTTTGACGTGGAGCTG | esiRNA cDNA target sequence GCATTATCCGGAGAACCAAACGGAAAGGAGAGGGATATTCAAATCCAAACGAAGGAGCAACAGTAGAAATCCACCTGGAAGGCCGCTGTGGTGGAAGGATGTTTGACTGCAGAGATGTGGCATTCACTGTGGGCGAAGGAGAAGACCACGACATTCCAATTGGAATTGACAAAGCTCTGGAGAAAATGCAGCGGGAAGAACAATGTATTTTATATCTTGGACCAAGATATGGTTTTGGAGAGGCAGGGAAGCCTAAATTTGGCATTGAACCTAATGCTGAGCTTATATATGAAGTTACACTTAAGAGCTTCGAAAAGGCCAAAGAATCCTGGGAGATGGATACCAAAGAAAAATTGGAGCAGGCTGCCATTGTCAAAGAGAAGGGAACCGTATACTTCAAGGGAGGCAAATACATGCAGGCGGTGAT | esiRNA cDNA target sequence GGACCACTGTCCCATCAAATCGCGCAAAGGGGATGTCCTGCACATGCACTACACGGGGAAGCTGGAAGATGGGACAGAGTTTGACAGCAGCCTGCCCCAGAACCAGCCCTTTGTCTTCTCCCTTGGCACAGGCCAGGTCATCAAGGGCTGGGACCAGGGGCTGCTGGGGATGTGTGAGGGGGAAAAGCGCAAGCTGGTGATCCCATCCGAGCTAGGGTATGGAGAGCGGGGAGCTCCCCCAAAGATTCCAGGTGGTGCAACCCTGGTGTTCGAGGTGGAGCTGCTCAAAATAGAGCGACGAACTGAGCTGTAACCAGACTGGGGAGGGGCAGGGGGAGAGGCCCCCATCAGGGACCAGACTGTTCCAAAAAAAAAACAAAAAACAAAAACAAACAAAAAAACACTTAAAAGCCCAAGGAGTAAGCC |
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| Gene Information human ... FKBP1A(2280) | Gene Information human ... FKBP1B(2281) | Gene Information human ... FKBP5(2289) | Gene Information human ... FKBP2(114841035), FKBP2(2286) |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
