Skip to Content
Merck
All Photos(1)

Key Documents

EHU068301

Sigma-Aldrich

MISSION® esiRNA

targeting human MAP3K5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTACCACCTTGGGGTGAGAGAAAGTTTCAGCATGGCCAACAACATCATCCTCTACTGTGATACTAACTCGGACTCTCTGCAGTCACTGAAGGAAATAATTTGCCAGAAGAATACTATGTGCACTGGGAACTACACCTTTGTTCCTTACATGATAACTCCACATAACAAAGTCTACTGCTGTGACAGCAGCTTCATGAAGGGGTTGACAGAGCTCATGCAACCGAACTTCGAGCTGCTTCTTGGACCCATCTGCTTACCTCTTGTGGATCGTTTTATTCAACTTTTGAAGGTGGCACAAGCAAGTTCTAGCCAGTACTTCCGGGAATCTATACTCAATGACATCAGGAAAGCTCGTAATTTATACACTGGTAAAGAATTGGCAGCTGAGTTGGCAAGAATTCGGCAGCGAGTAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongwei Yao et al.
Biochemical and biophysical research communications, 493(3), 1288-1295 (2017-10-03)
Interleukin-33 (IL-33), a new member of the IL-1 cytokine family, has cardiac protective effect in many circumstances. The aims of present study are to assess whether IL-33 can protect cardiomyocytes from doxorubicin (DOX)-induced apoptosis and the mechanism involved in the
Liwei Ma et al.
Journal of experimental & clinical cancer research : CR, 38(1), 77-77 (2019-02-15)
Metformin, a first-line drug for type 2 diabetes, could induce apoptosis in cancer cells. However, the concentration of glucose affects the effect of metformin, especially low glucose in the culture medium can enhance the cytotoxicity of metformin on cancer cells.
Chiara Imarisio et al.
Free radical biology & medicine, 112, 141-148 (2017-07-26)
Steatosis intensifies hepatic ischemia/reperfusion (I/R) injury increasing hepatocyte damage and hepatic inflammation. This study evaluates if this process is associated to a differential response of steatotic hepatocytes (HP) and Kupffer cells (KC) to I/R injury and investigates the molecular mechanisms
Mathew S Eapen et al.
Clinical science (London, England : 1979), 132(14), 1615-1627 (2018-07-15)
Increased airway smooth muscle (ASM) mass is observed in chronic obstructive pulmonary disease (COPD), which is correlated with disease severity and negatively affects lung function in these patients. Thus, there is clear unmet clinical need for finding new therapies which
Musarat Ishaq et al.
Biochimica et biophysica acta, 1843(12), 2827-2837 (2014-09-01)
Atmospheric pressure gas plasma (AGP) generates reactive oxygen species (ROS) that induce apoptosis in cultured cancer cells. The majority of cancer cells develop a ROS-scavenging anti-oxidant system regulated by Nrf2, which confers resistance to ROS-mediated cancer cell death. Generation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service