Skip to Content
Merck

Skip To

EHU034671

MISSION® esiRNA

targeting human PIK3CB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
CZK 5,620.00
50 μg
Please contact Customer Service for Availability
CZK 10,100.00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

CZK 5,620.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAAGCCTTTGTGGCTGGTATACAATAACAAGGTATTTGGTGAGGATTCAGTTGGAGTGATTTTTAAAAATGGTGATGATTTACGACAGGATATGTTGACACTCCAAATGTTGCGCTTGATGGATTTACTCTGGAAAGAAGCTGGTTTGGATCTTCGGATGTTGCCTTATGGCTGTTTAGCAACAGGAGATCGCTCTGGCCTCATTGAAGTTGTGAGCACCTCTGAAACAATTGCTGACATTCAGCTGAACAGTAGCAATGTGGCTGCTGCAGCAGCCTTCAACAAAGATGCCCTTCTGAACTGGCTTAAAGAATACAACTCTGGGGATGACCTGGACCGAGCCATTGAGGAATTTACACTGTCCTGTGCTGGCTACTGTGTAGCTTCTTATGTCCTTGGGATTGGTGACAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... PIK3CB(5291)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHU113461EHU039391EHU042421
esiRNA cDNA target sequence

GAAGCCTTTGTGGCTGGTATACAATAACAAGGTATTTGGTGAGGATTCAGTTGGAGTGATTTTTAAAAATGGTGATGATTTACGACAGGATATGTTGACACTCCAAATGTTGCGCTTGATGGATTTACTCTGGAAAGAAGCTGGTTTGGATCTTCGGATGTTGCCTTATGGCTGTTTAGCAACAGGAGATCGCTCTGGCCTCATTGAAGTTGTGAGCACCTCTGAAACAATTGCTGACATTCAGCTGAACAGTAGCAATGTGGCTGCTGCAGCAGCCTTCAACAAAGATGCCCTTCTGAACTGGCTTAAAGAATACAACTCTGGGGATGACCTGGACCGAGCCATTGAGGAATTTACACTGTCCTGTGCTGGCTACTGTGTAGCTTCTTATGTCCTTGGGATTGGTGACAG

esiRNA cDNA target sequence

CTGGCTGAAGTCCAAGAACCCGGGGGAGGCCCTGGATCGAGCCATTGAGGAGTTCACCCTCTCCTGTGCTGGCTATTGTGTGGCCACATATGTGCTGGGCATTGGCGATCGGCACAGCGACAACATCATGATCCGAGAGAGTGGGCAGCTGTTCCACATTGATTTTGGCCACTTTCTGGGGAATTTCAAGACCAAGTTTGGAATCAACCGCGAGCGTGTCCCATTCATCCTCACCTACGACTTTGTCCATGTGATTCAGCAGGGGAAGACTAATAATAGTGAGAAATTTGAACGGTTCCGGGGCTACTGTGAAAGGGCCTACACCATCCTGCGGCGCCACGGGCTTCTCTTCCTCCACCTCTTTGCCCTGATGCGGGCGGCAGGCCTGCCTGAGCTCAGCTGCTCCAAAGACATC

esiRNA cDNA target sequence

ACCTTGTTCCAATCCCAGGTGGAATGAATGGCTGAATTATGATATATACATTCCTGATCTTCCTCGTGCTGCTCGACTTTGCCTTTCCATTTGCTCTGTTAAAGGCCGAAAGGGTGCTAAAGAGGAACACTGTCCATTGGCATGGGGAAATATAAACTTGTTTGATTACACAGACACTCTAGTATCTGGAAAAATGGCTTTGAATCTTTGGCCAGTACCTCATGGATTAGAAGATTTGCTGAACCCTATTGGTGTTACTGGATCAAATCCAAATAAAGAAACTCCATGCTTAGAGTTGGAGTTTGACTGGTTCAGCAGTGTGGTAAAGTTCCCAGATATGTCAGTGATTGAAGAGCATGCCAATTGGTCTGTATCCCGAGAAGCAGGATTTAGCTATTCCCACGCAGG

esiRNA cDNA target sequence

TCAGCCAAGCATTGTTGAAGGGTGATAAGTCTGTCAGAGTTATGCGTTCTTTGCTGGCTGCACAACAGACATTTGTAGATCGGTTGGTGCATCTAATGAAGGCAGTACAACGCGAAAGTGGAAATCGTAAGAAAAAGAATGAGAGACTACAGGCATTGCTTGGAGATAATGAAAAGATGAATTTGTCAGATGTGGAACTTATCCCGTTGCCTTTAGAACCCCAAGTGAAAATTAGAGGAATAATTCCGGAAACAGCTACACTGTTTAAAAGTGCCCTTATGCCTGCACAGTTGTTTTTTAAGACGGAAGATGGAGGCAAATATCCAGTTATATTTAAGCATGGAGATGATTTACGTCAAGATCAACTTATTCTTCAAATCATTTCACTCATGGACAAGCTGTTACGG

Gene Information

human ... PIK3CB(5291)

Gene Information

human ... PIK3CD(5293)

Gene Information

human ... PIK3CA(5290)

Gene Information

human ... PIK3C3(5289)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | human accession no.

ENSG00000051382

Ensembl | human accession no.

ENSG00000171608

Ensembl | human accession no.

ENSG00000121879

Ensembl | human accession no.

ENSG00000078142

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

NM_006219

NCBI accession no.

NM_005026

NCBI accession no.

NM_006218

NCBI accession no.

NM_002647


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters