Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 20 μg | Please contact Customer Service for Availability | CZK 5,620.00 | |
| 50 μg | Please contact Customer Service for Availability | CZK 10,100.00 |
About This Item
CZK 5,620.00
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GAGCAGAACTTCGTGTGCAAGTTCGCGCCCAGGGAGGGCTTCATCAACTACCTCACGAGGGAAGTGTACCGCTCCTACCGCCAGCTGCGGACCCAGGGCTTTGGAGGGTCTGGGATCCCCAAGGCCTGGGCAGGCATAGACTTGAAGGTACAACCCCAGGAACCCCTGGTGCTGAAGGATGTGGAAAACACAGATTGGCGCCTACTGCGGGGTGACACGGATGTCAGGGTAGAGAGGAAAGACCCAAACCAGGTGGAACTGTGGGGACTCAAGGAAGGCACCTACCTGTTCCAGCTGACAGTGACTAGCTCAGACCACCCAGAGGACACGGCCAACGTCACAGTCACTGTGCTGTCCACCAAGCAGACAGAAGACTACTGCCTCGCATCCAACAAGGTGGGTCGCTGCCGGGGCTCTTTCCCACGCTGGTACTATGACCCCACGGAGCAGATCTGCAAGAGTTTCGTTTATGGAGGCTGCT
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... SPINT1(6692)
1 of 1
This Item | |||
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| esiRNA cDNA target sequence GAGCAGAACTTCGTGTGCAAGTTCGCGCCCAGGGAGGGCTTCATCAACTACCTCACGAGGGAAGTGTACCGCTCCTACCGCCAGCTGCGGACCCAGGGCTTTGGAGGGTCTGGGATCCCCAAGGCCTGGGCAGGCATAGACTTGAAGGTACAACCCCAGGAACCCCTGGTGCTGAAGGATGTGGAAAACACAGATTGGCGCCTACTGCGGGGTGACACGGATGTCAGGGTAGAGAGGAAAGACCCAAACCAGGTGGAACTGTGGGGACTCAAGGAAGGCACCTACCTGTTCCAGCTGACAGTGACTAGCTCAGACCACCCAGAGGACACGGCCAACGTCACAGTCACTGTGCTGTCCACCAAGCAGACAGAAGACTACTGCCTCGCATCCAACAAGGTGGGTCGCTGCCGGGGCTCTTTCCCACGCTGGTACTATGACCCCACGGAGCAGATCTGCAAGAGTTTCGTTTATGGAGGCTGCT | esiRNA cDNA target sequence GCCTCAAGAAATGTGCCACTGTCACAGAGAATGCCACGGGTGACCTGGCCACCAGCAGGAATGCAGCGGATTCCTCTGTCCCAAGTGCTCCCAGAAGGCAGGATTCTGAAGACCACTCCAGCGATATGTTCAACTATGAAGAATACTGCACCGCCAACGCAGTCACTGGGCCTTGCCGTGCATCCTTCCCACGCTGGTACTTTGACGTGGAGAGGAACTCCTGCAATAACTTCATCTATGGAGGCTGCCGGGGCAATAAGAACAGCTACCGCTCTGAGGAGGCCTGCATGCTCCGCTGCTTCCGCCAGCAGGAGAATCCTCCCCTGCCCCTTGGCTCAAAGGTGGTGGTTCTGGCGGGGCTGTTCGTGATGGTGTTGATCCTCTTCCTGGGAGCCTCCATGGTCTACCTGATCCGGGTGGCACGGAGGAACCAGGAGCGTGCCCTGCGCACCGTCTGGAGCTCCGGAGATGACAAGGAGCAGCTGGTGAAGAA | esiRNA cDNA target sequence CTTCACACCAGGACCAGTCACTGTTGCAGCATGAGTTCCCAGCAGCAGAAGCAGCCTTGCACCCCACCCCCTCAGCTTCAGCAGCAGCAGGTGAAACAGCCTTGCCAGCCTCCACCTCAGGAACCATGCATCCCCAAAACCAAGGAGCCCTGCCACCCCAAGGTGCCTGAGCCCTGCCACCCCAAAGTGCCCGAGCCCTGCCAGCCCAAGGTTCCAGAGCCATGCCACCCCAAGGTGCCTGAGCCCTGCCCTTCAATAGTCACTCCAGCACCAGCCCAGCAGAAGACCAAGCAGAAGTAATGTGGTCCACAGCCATGCCCTTGAGGAGCCGGCCACCAGATGCTGAATCCCCTATCCCATTCTGCGTATGAGTCCCATTTGCCTTGCAATTAGCATTCTGTCTCCCCCAAAAA | esiRNA cDNA target sequence TGCCATATGACCCTTCCAGTCCCAGGCTTCTGAAGAGACGTGGTAAGTGCGGTGCAGTTTTCAACTGACCTCTGGACGCAGAACTTCAGCCATGAAGGTAACAGGCATCTTTCTTCTCAGTGCCTTGGCCCTGTTGAGTCTATCTGGTAACACTGGAGCTGACTCCCTGGGAAGAGAGGCCAAATGTTACAATGAACTTAATGGATGCACCAAGATATATGACCCTGTCTGTGGGACTGATGGAAATACTTATCCCAATGAATGCGTGTTATGTTTTGAAAATCGGAAACGCCAGACTTCTATCCTCATTCAAAAATCTGGGCCTTGCTGAGAACCAAGGTTTTGAAATCCCATCAGGTCACC |
| Gene Information human ... SPINT1(6692), SPINT1(6692) | Gene Information human ... SPINT2(10653), SPINT2(10653) | Gene Information human ... SPRR1B(6699), SPRR1B(6699) | Gene Information human ... SPINK1(6690), SPINK1(6690) |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EHU030331-50UG | 04061828344079 |
| EHU030331-20UG | 04061828579150 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service