Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C12H12N5O7PNa2
Molecular Weight:
415.21
PubChem Substance ID:
UNSPSC Code:
41106305
form
solid
storage temp.
−20°C
SMILES string
O[C@H]([C@@H](COP(O)(O)=O)O1)[C@@H](O)[C@@H]1N(C=N2)C3=C2C4=NC=CN4C=N3.[Na]
InChI
1S/C12H14N5O7P.Na.H/c18-8-6(3-23-25(20,21)22)24-12(9(8)19)17-5-14-7-10-13-1-2-16(10)4-15-11(7)17;;/h1-2,4-6,8-9,12,18-19H,3H2,(H2,20,21,22);;
InChI key
PFRNHZIHKGVNJU-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.