Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H34O2
CAS Number:
Molecular Weight:
306.48
UNSPSC Code:
12352211
PubChem Substance ID:
EC Number:
200-578-6
MDL number:
assay
≥90%
form
liquid
concentration
~10 mg/mL in ethanol
functional group
carboxylic acid
storage temp.
−20°C
SMILES string
CCCCCCCC\C=C/C\C=C/C\C=C/CCCC(O)=O
InChI
1S/C20H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h9-10,12-13,15-16H,2-8,11,14,17-19H2,1H3,(H,21,22)/b10-9-,13-12-,16-15-
InChI key
UNSRRHDPHVZAHH-YOILPLPUSA-N
Packaging
Packaged under Argon.
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
57.2 °F - closed cup - Solvent
flash_point_c
14.0 °C - closed cup - Solvent
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

