Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C21H40O2
CAS Number:
Molecular Weight:
324.54
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
assay
~99% (capillary GC)
form
liquid
functional group
ester
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCCC\C=C/CCCCCCCCCCCC(=O)OC
InChI
1S/C21H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h8-9H,3-7,10-20H2,1-2H3/b9-8-
InChI key
XQXARCJTXXEQEX-HJWRWDBZSA-N
Packaging
Sealed ampule.
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.