Skip to Content
Merck

Skip To

E1656

Endocleaning Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

room temp

Quality Segment

General description

For sample preparation use in some GenElute kits.

Preparation Note

For usage requirements, please refer to the kit procedure.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B9560N7411Q2259
description

-

description

-

description

-

description

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters