Skip to Content
Merck
All Photos(1)

Key Documents

E1158

Sigma-Aldrich

EPHB3 (585-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Cek10, ETK2, HEK2, Mdk5, TYRO6

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 9,380.00

CZK 9,380.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
CZK 9,380.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CZK 9,380.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

22-30 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EPHB3(2049)

Biochem/physiol Actions

EPHB3 is a member of the Ephrin receptor family and is expressed during embryonic development in multiple regions of the central nervous system. In adult brain, EPHB3 is expressed in the cerebellum, raphe pallidus, hippocampus, entorhinal cortex, and both motor and sensory cortices. EPHB3 is involved in the maintenance of mature neuronal connections and/or re-arrangement of synaptic connections during late stages of development. EPHB3 plays a role in the regulation of cell adhesion and migration, and the catalytic activity of EPHB3 is required for inhibition of integrin-mediated cell adhesion.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher A Willson et al.
Journal of molecular histology, 37(8-9), 369-380 (2006-11-15)
Eph receptors and ligands are two families of proteins that control axonal guidance during development. Their expression was originally thought to be developmentally regulated but recent work has shown that several EphA receptors are expressed postnatally. The EphB3 receptors are
Hui Miao et al.
The Journal of biological chemistry, 280(2), 923-932 (2004-11-13)
Genetic studies have shown that Eph receptor tyrosine kinases have both kinase-dependent and kinase-independent functions through incompletely understood mechanisms. We report here that ephrin-B1 stimulation of endogenous EphB kinases in LS174T colorectal epithelial cells inhibited integrin-mediated adhesion and HGF/SF-induced directional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service