Skip to Content
Merck
All Photos(1)

Key Documents

E1151

Sigma-Aldrich

Anti-ERAB antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Endoplasmic Reticulum-Associated Amyloid β-Peptide-Binding Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1-2 μg/mL using human cerebellum tissue
western blot: 1-3 μg/mL using human SK-N-SH cells

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... HSD17B10(3028)

General description

The gene HSD17B10 (hydroxysteroid (17-β) dehydrogenase 10) encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily.

Specificity

By immunoblotting, the antibody detects native human ERAB. It does not react with mouse or rat.

Immunogen

synthetic peptide corresponding to amino acids 100-116 of human ERAB protein.

Application

Anti-ERAB antibody produced in rabbit is suitable for immunohistochemistry at a working concentration of 1-2μg/mL using human cerebellum tissue with DAB detection and for immunoblotting at 1-3μg/mL using human neuroblastoma SK-N-SH cells with chemiluminescence detection.

Biochem/physiol Actions

The gene product is a subunit of mitochondrial ribonuclease P and plays a pivotal role in in mitochondrial tRNA maturation. It catalyzes the oxidation of a wide variety of fatty acids and steroids. The gene is linked to the neuronal dysfunction associated with Alzheimer disease (AD).

Physical form

Solution in 0.05 M sodium phosphate buffer, pH 7.5, containing 0.2% gelatin and 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Yan et al.
Nature, 389(6652), 689-695 (1997-10-24)
Amyloid-beta is a neurotoxic peptide which is implicated in the pathogenesis of Alzheimer's disease. It binds an intracellular polypeptide known as ERAB, thought to be a hydroxysteroid dehydrogenase enzyme, which is expressed in normal tissues, but is overexpressed in neurons
Naeem Shafqat et al.
The Biochemical journal, 376(Pt 1), 49-60 (2003-08-15)
17beta-hydroxysteroid dehydrogenases (17beta-HSDs) catalyse the conversion of 17beta-OH (-hydroxy)/17-oxo groups of steroids, and are essential in mammalian hormone physiology. At present, eleven 17beta-HSD isoforms have been defined in mammals, with different tissue-expression and substrate-conversion patterns. We analysed 17beta-HSD type 10
Johann Holzmann et al.
Cell, 135(3), 462-474 (2008-11-06)
tRNAs are synthesized as immature precursors, and on their way to functional maturity, extra nucleotides at their 5' ends are removed by an endonuclease called RNase P. All RNase P enzymes characterized so far are composed of an RNA plus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service