Skip to Content
Merck
All Photos(1)

Documents

D9925

Sigma-Aldrich

1,2-Dioleoyl-3-linoleoyl-rac-glycerol

liquid, 97% (TLC)

Synonym(s):

1,2-Di(cis-9-octadecenoyl)-3-(cis-9,12-octadecadienoyl)-rac-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H102O6
CAS Number:
Molecular Weight:
883.42
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

97% (TLC)

form

liquid

functional group

ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\C\C=C\CCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C57H102O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16,19,25-30,54H,4-15,17-18,20-24,31-53H2,1-3H3/b19-16+,28-25+,29-26+,30-27+

InChI key

JTMWOTXEVWLTTO-CGFQLDJTSA-N

Physical form

Viscous liquid at room temperature.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service