Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Linear Formula:
(CH3O)2C6H3CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
EC Number:
244-036-7
UNSPSC Code:
12352103
MDL number:
mp
188-190 °C (lit.)
storage temp.
−20°C
SMILES string
Cl.COc1ccc(CN)c(OC)c1
InChI
1S/C9H13NO2.ClH/c1-11-8-4-3-7(6-10)9(5-8)12-2;/h3-5H,6,10H2,1-2H3;1H
InChI key
KJOWSAVFSCSBMZ-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service