Skip to Content

D8886

Anti-Dehydroepiandrosterone (DHEA) antibody produced in rabbit

whole antiserum, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

radioimmunoassay: suitable

storage temp.

2-8°C

Immunogen

DHEA-15α-CH2-CO-BSA.

Biochem/physiol Actions

Dehydroepiandrosterone is a sulfated neurosteroid, abundantly present in the brain. It is a strong allosteric modulator of GABAA receptors and has antiseizure and anesthetic properties.

Preparation Note

Reconstitute with 11 ml 0.1 M phosphate buffered saline, pH 7.4, containing 0.1 M gelatin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library