Skip to Content
Merck

D8275

Dicyandiamide

Synonym(s):

Cyanoguanidine, Dicyanodiamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2C(=NH)NHCN
CAS Number:
Molecular Weight:
84.08
EC Number:
207-312-8
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
605637
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

208-211 °C (lit.)

storage temp.

2-8°C

SMILES string

NC(=N)NC#N

InChI

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

InChI key

QGBSISYHAICWAH-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library