Skip to Content
Merck

D8134

4-(Dimethylamino)benzylamine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2NC6H4CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
223.14
UNSPSC Code:
12352103
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

220-224 °C (dec.) (lit.)

SMILES string

Cl.Cl.CN(C)c1ccc(CN)cc1

InChI

1S/C9H14N2.2ClH/c1-11(2)9-5-3-8(7-10)4-6-9;;/h3-6H,7,10H2,1-2H3;2*1H

InChI key

NRZQHDOWSPJUQW-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library