Skip to Content
Merck
All Photos(1)

Key Documents

D7690

Sigma-Aldrich

Monoclonal Anti-DNA Ligase I antibody produced in mouse

~1 mg/mL, clone 1A9, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~125 kDa

species reactivity

bovine, human

concentration

~1 mg/mL

technique(s)

direct ELISA: suitable
western blot: 1-5 μg/mL using HeLa or DiFi cells

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIG1(3978)

General description

DNA ligase I is composed of a 78kDa carboxyl-terminal catalytic domain and a 24kDa amino-terminal region.

Immunogen

bovine DNA ligase I.

Application

Monoclonal Anti-DNA Ligase I antibody produced in mouse is suitable for direct ELISA and western blot at a working concentration of 1-5μg/mL using HeLa or DiFi cells.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V J Mackenney et al.
The Journal of biological chemistry, 272(17), 11550-11556 (1997-04-25)
The joining of Okazaki fragments during lagging strand DNA replication in mammalian cells is believed to be due to DNA ligase I. This enzyme is composed of a 78-kDa carboxyl-terminal catalytic domain and a 24-kDa amino-terminal region that is not

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service